Learn More
4-Chloro-2-fluoro-N-methylbenzamide, TRC
Product Code. 30366251
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366251 100 mg 100mg
30361027 500 mg -
30355830 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30366251 Supplier Toronto Research Chemicals Supplier No. TRCC584805100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1343038-33-4
Molecular Formula C8 H7 Cl F N O
Molecular Weight (g/mol) 187.599
Synonym Apalutamide Impurity 27
IUPAC Name 4-chloro-2-fluoro-N-methylbenzamide
SMILES CNC(=O)c1ccc(Cl)cc1F
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C8H7ClFNO/c1-11-8(12)6-3-2-5(9)4-7(6)10/h2-4H,1H3,(H,11,12)
Formula Weight 187.02
Chemical Name or Material 4-Chloro-2-fluoro-N-methylbenzamide