accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CH3CCH3OOOCH3H3CH3C

CAS RN 13402-96-5

IUPAC Name: 2-[2,4-bis(2-methylbutan-2-yl)phenoxy]acetate
Synonyms: 2-[2,4-bis(2-methylbutan-2-yl)phenoxy]acetate
Molecular Weight (g/mol): 291.41
Molecular Formula: C18H27O3
InChi Key: QXQMENSTZKYZCE-UHFFFAOYSA-M
SMILES: CCC(C)(C)C1=CC=C(OCC([O-])=O)C(=C1)C(C)(C)CC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results