missing translation for 'onlineSavingsMsg'
Learn More

2,4-Di(tert-pentyl)phenoxyacetic acid, 98%, Thermo Scientific™

Product Code. 11428477
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428477 500 g 500g
11408477 25 g 25g
11418477 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428477 Supplier Thermo Scientific Alfa Aesar Supplier No. A18546.36
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13402-96-5
Molecular Formula C18H27O3
Molecular Weight (g/mol) 291.41
MDL Number MFCD00128799
InChI Key QXQMENSTZKYZCE-UHFFFAOYSA-M
Synonym 2-2,4-di-tert-pentylphenoxy acetic acid, 2,4-di tert-amyl phenoxyacetic acid, 2,4-di-tert-pentylphenoxy acetic acid, 2,4-di tert-pentyl phenoxyacetic acid, 2,4-di-tert-pentylphenoxyacetic acid, 2-2,4-bis 2-methylbutan-2-yl phenoxy acetic acid, acetic acid, 2,4-bis 1,1-dimethylpropyl phenoxy, 2,4-bis 1,1-dimethylpropyl phenoxy acetic acid, acetic acid, 2-2,4-bis 1,1-dimethylpropyl phenoxy
PubChem CID 83411
IUPAC Name 2-[2,4-bis(2-methylbutan-2-yl)phenoxy]acetic acid
SMILES CCC(C)(C)C1=CC=C(OCC([O-])=O)C(=C1)C(C)(C)CC

Specifications

Melting Point 125°C to 127°C
Quantity 500 g
Formula Weight 292.42
Percent Purity 98%
Chemical Name or Material 2,4-Di(tert-pentyl)phenoxyacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.