CAS RN 132682-23-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-4-(Hydroxymethyl)oxazolidin-2-one, TRC
CAS: 132682-23-6 Molecular Formula: C4H7NO3 Molecular Weight (g/mol): 117.1 Synonym: (4R)-4-(Hydroxymethyl)-1,3-oxazolidin-2-one,(4R)-4-(Hydroxymethyl)-2-oxazolidinone IUPAC Name: (4R)-4-(hydroxymethyl)-1,3-oxazolidin-2-one SMILES: OC[C@@H]1COC(=O)N1