Learn More
(R)-4-(Hydroxymethyl)oxazolidin-2-one, TRC
Product Code. 17992725
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17992725 2.5 g 2.5g
17932535 1 g 1g
17972065 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17992725 Supplier Toronto Research Chemicals Supplier No. TRCH8053002.5G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 132682-23-6
Molecular Formula C4H7NO3
Molecular Weight (g/mol) 117.1
Synonym (4R)-4-(Hydroxymethyl)-1,3-oxazolidin-2-one, (4R)-4-(Hydroxymethyl)-2-oxazolidinone
IUPAC Name (4R)-4-(hydroxymethyl)-1,3-oxazolidin-2-one
SMILES OC[C@@H]1COC(=O)N1
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C4H7NO3/c6-1-3-2-8-4(7)5-3/h3,6H,1-2H2,(H,5,7)/t3-/m1/s1
Formula Weight 117.0426
Chemical Name or Material (R)-4-(Hydroxymethyl)oxazolidin-2-one