CAS RN 13080-75-6
CAS RN 13080-75-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Acetyl-10,11-dihydro-5H-dibenzo[b,f]azepine (N-Acetyliminodibenzyl), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.