Learn More
N-Acetyl-10,11-dihydro-5H-dibenzo[b,f]azepine (N-Acetyliminodibenzyl), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
N-Acetyl-10,11-dihydro-5H-dibenzo[b,f]azepine (N-Acetyliminodibenzyl)
Specifications
Specifications
| Chemical Name or Material | N-Acetyliminodibenzyl |
| CAS | 13080-75-6 |
| Analyte or Component Names | N-Acetyliminodibenzyl |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H15 N O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-(10,11-Dihydro-5H-dibenz[b,f]azepin-5-yl)ethanone, 5H-Dibenz[b,f]azepine, 5-acetyl-10,11-dihydro- (6CI,7CI,8CI,9CI), 1-(10,11-Dihydro-5H-dibenz[b,f]azepin-5-yl)ethanone, 1-(10,11-Dihydrodibenz[b,f]azepin-5-yl)ethanone, 5-Acetyl-10,11-dihydro-5H-dibenz[b,f]azepine, 5-Acetyl-10,11-dihydro-5H-dibenzo[b,f]azepine, N-Acetyl-10,11-dihydro-5H-dibenzo[b,f]azepine, N-Acetyliminodibenzyl |
| InChI Formula | InChI=1S/C16H15NO/c1-12(18)17-15-8-4-2-6-13(15)10-11-14-7-3-5-9-16(14)17/h2-9H,10-11H2,1H3 |
| SMILES | CC(=O)N1c2ccccc2CCc3ccccc13 |
| IUPAC Name | 1-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)ethanone |
| Show More |