accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NBrN

CAS RN 1260805-87-5

IUPAC Name: 2-(azepan-1-yl)-6-bromobenzonitrile
Synonyms: 2-(azepan-1-yl)-6-bromobenzonitrile
Molecular Weight (g/mol): 279.18
Molecular Formula: C13H15BrN2
InChi Key: PFOLTHPOEJJYAZ-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(N2CCCCCC2)=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results