Learn More
2-(1-Azepanyl)-6-bromobenzonitrile, 96%, Thermo Scientific™
Product Code. 15487718
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487718 250 mg 250mg
15477718 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15487718 Supplier Thermo Scientific Alfa Aesar Supplier No. H61216.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1260805-87-5
Molecular Formula C13H15BrN2
Molecular Weight (g/mol) 279.181
InChI Key PFOLTHPOEJJYAZ-UHFFFAOYSA-N
Synonym 2-azepan-1-yl-6-bromobenzonitrile
PubChem CID 75360876
IUPAC Name 2-(azepan-1-yl)-6-bromobenzonitrile
SMILES C1CCCN(CC1)C2=C(C(=CC=C2)Br)C#N
Specifications
Quantity 250 mg
UN Number UN3439
Formula Weight 279.18
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-(1-Azepanyl)-6-bromobenzonitrile