CAS RN 1189426-16-1
CAS RN 1189426-16-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Sulfadiazine-13C6, TRC
CAS: 1189426-16-1 Molecular Formula: 13C6 C4 H10 N4 O2 S Molecular Weight (g/mol): 256.23 Synonym: Benzene-1,2,3,4,5,6-13C6-sulfonamide, 4-amino-N-2-pyrimidinyl- IUPAC Name: 4-amino-N-pyrimidin-2-yl(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene-1-sulfonamide SMILES: N[13c]1[13cH][13cH][13c]([13cH][13cH]1)S(=O)(=O)Nc2ncccn2
2
Sulfadiazine 13C6 (phenyl 13C6) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality