Learn More
Sulfadiazine 13C6 (phenyl 13C6) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Sulfadiazine-13C6 |
| CAS | 1189426-16-1 |
| Solvent or Matrix | Acetonitrile |
| Molecular Formula | 13C6 C4 H10 N4 O2 S |
| Quantity | 1 mL |
| Synonym | Benzene-1,2,3,4,5,6-13C6-sulfonamide, 4-amino-N-2-pyrimidinyl- |
| InChI Formula | InChI=1S/C10H10N4O2S/c11-8-2-4-9(5-3-8)17(15,16)14-10-12-6-1-7-13-10/h1-7H,11H2,(H,12,13,14)/i2+1,3+1,4+1,5+1,8+1,9+1 |
| SMILES | N[13c]1[13cH][13cH][13c]([13cH][13cH]1)S(=O)(=O)Nc2ncccn2 |
| IUPAC Name | 4-amino-N-pyrimidin-2-yl(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene-1-sulfonamide |
| Molecular Weight (g/mol) | 256.23 |
| Show More |