Learn More
Sulfadiazine-13C6, TRC
Product Code. 30010306
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010306 1 mg 1mg
30008175 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30010306 Supplier Toronto Research Chemicals Supplier No. TRCS6990521MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1189426-16-1
Molecular Formula 13C6 C4 H10 N4 O2 S
Molecular Weight (g/mol) 256.23
Synonym Benzene-1,2,3,4,5,6-13C6-sulfonamide, 4-amino-N-2-pyrimidinyl-
IUPAC Name 4-amino-N-pyrimidin-2-yl(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene-1-sulfonamide
SMILES N[13c]1[13cH][13cH][13c]([13cH][13cH]1)S(=O)(=O)Nc2ncccn2
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C10H10N4O2S/c11-8-2-4-9(5-3-8)17(15,16)14-10-12-6-1-7-13-10/h1-7H,11H2,(H,12,13,14)/i2+1,3+1,4+1,5+1,8+1,9+1
Formula Weight 256.0726
Percent Purity >95%
Chemical Name or Material Sulfadiazine-13C6
Grade HPLC