Specifications
| Quantity | 100 mg |
| Solubility | Solubility aq. soln.: .. Other solubilities: soluble in dmso, insoluble in methanol |
| Melting Point | 249.0°C to 251.0°C |
| Formula Weight | 448.58 |
| InChI Key | NSYUKKYYVFVMST-LETVYOFWSA-L |
| SMILES | [Ca++].C[C@H](CSC(=O)C1=CC=CC=C1)C(=O)N1C[C@H](C[C@H]1C([O-])=O)SC1=CC=CC=C1.C[C@H](CSC(=O)C1=CC=CC=C1)C(=O)N1C[C@H](C[C@H]1C([O-])=O)SC1=CC=CC=C1 |
| IUPAC Name | calcium bis((2S,4S)-1-[(2S)-3-(benzoylsulfanyl)-2-methylpropanoyl]-4-(phenylsulfanyl)pyrrolidine-2-carboxylate) |
| Molecular Weight (g/mol) | 897.16 |
| Chemical Name or Material | Zofenopril calcium |