accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 81938-43-4

CaCH3(S)SOON(S)(S)OOSCH3(S)SOON(S)(S)OOS

CAS RN 81938-43-4

IUPAC Name: calcium bis((2S,4S)-1-[(2S)-3-(benzoylsulfanyl)-2-methylpropanoyl]-4-(phenylsulfanyl)pyrrolidine-2-carboxylate)
Synonyms: calcium bis((2S,4S)-1-[(2S)-3-(benzoylsulfanyl)-2-methylpropanoyl]-4-(phenylsulfanyl)pyrrolidine-2-carboxylate)
Molecular Weight (g/mol): 897.16
Molecular Formula: C44H44CaN2O8S4
InChi Key: NSYUKKYYVFVMST-LETVYOFWSA-L
SMILES: [Ca++].C[C@H](CSC(=O)C1=CC=CC=C1)C(=O)N1C[C@H](C[C@H]1C([O-])=O)SC1=CC=CC=C1.C[C@H](CSC(=O)C1=CC=CC=C1)C(=O)N1C[C@H](C[C@H]1C([O-])=O)SC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results