Learn More
Zaleplon, Mikromol™
Product Code. 17991616
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 17991616
Brand: Mikromol MM1322.00
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Zaleplon
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Zaleplon
CAS 151319-34-5
Analyte or Component Names Zaleplon
Shipping Condition Room Temperature
Molecular Formula C17 H15 N5 O
Certifications/Compliance ISO 17034
Synonym Zaleplon
InChI Formula InChI=1S/C17H15N5O/c1-3-21(12(2)23)15-6-4-5-13(9-15)16-7-8-19-17-14(10-18)11-20-22(16)17/h4-9,11H,3H2,1-2H3
SMILES CCN(C(=O)C)c1cccc(c1)c2ccnc3c(cnn23)C#N
IUPAC Name N-[3-(3-cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]-N-ethylacetamide
Molecular Weight (g/mol) 305.33
Quantity 100 mg
Formula Weight 305.1277 g/mol