Learn More
Yohimbine Hydrochloride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Yohimbine hydrochloride |
| CAS | 65-19-0 |
| Analyte or Component Names | Yohimbine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H26 N2 O3 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Yohimbine hydrochloride, Benz[g]indolo[2,3-a]quinolizine, yohimban-16-carboxylic acid deriv., Actibine, Antagonil, Aphrodyne, Dayto-Himbin, Erex, NIH 9689, NSC 19509, Thybine, Yocon, Yohimbe, Yohimbex, Yohimbine chloride, Yohimbine monohydrochloride, Yohimex, Yomax, Yohimban-16-carboxylic acid, 17-hydroxy-, methyl ester, monohydrochloride, (16alpha,17alpha)- (9CI), Yohimban-16alpha-carboxylic acid, 17alpha-hydroxy-, methyl ester, monohydrochloride (8CI) |
| InChI Formula | InChI=1S/C21H26N2O3.ClH/c1-26-21(25)19-15-10-17-20-14(13-4-2-3-5-16(13)22-20)8-9-23(17)11-12(15)6-7-18(19)24;/h2-5,12,15,17-19,22,24H,6-11H2,1H3;1H/t12-,15-,17-,18-,19+;/m0./s1 |
| Solution Type | Neat |
| SMILES | Cl.COC(=O)[C@H]1[C@@H](O)CC[C@H]2CN3CCc4c([nH]c5ccccc45)[C@@H]3C[C@H]12 |
| Show More |