Learn More
Waters Corporation XTerra MS C8 Column
XTerra MS C8 columns are general purpose, hybrid-based, reversed-phase C8 columns that are designed to be compatible with mass spectrometry applications.
Brand:  Waters Corporation 186000433
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 13222590
  • 1192.50 € / Each
Estimated Shipment: 21-06-2024
to see stock.
Add to basket
Description
Description
  • These first generation columns provide superior pH stability compared to a silica-based column
Specifications
Specifications
4.6 mm
6000 psi (415 Bar)
3.5 μm
1/Ea.
Yes
50 mm
Spherical
125 Å
L7
Column