Learn More
Xanthinol Nicotinate, Mikromol™
Description
Xanthinol Nicotinate
Specifications
Specifications
| Chemical Name or Material | Xanthinol Nicotinate |
| CAS | 437-74-1 |
| Analyte or Component Names | Xanthinol Nicotinate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H21 N5 O4 . C6 H5 N O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 3-Pyridinecarboxylic acid, compd. with 3,7-dihydro-7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-1,3-dimethyl-1H-purine-2,6-dione (1:1), Nicotinic acid, compd. with 7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]theophylline (1:1) (8CI), Theophylline, 7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-, nicotinate (6CI), 1H-Purine-2,6-dione, 3,7-dihydro-7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-1,3-dimethyl-, mono-3-pyridinecarboxylate (salt) (9CI), Theophylline, 7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-, mononicotinate (salt) (8CI), 7-[2-Hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]theophylline nicotinate, 7-[2-Hydroxy-3-[N-(2-hydroxyethyl)-N-methylamino]propyl]-1,3-dimethylxanthine pyridine-3-carboxylate, 7-[2-Hydroxy-3-[N-methyl-N-(2-hydroxyethyl)amino]-propyl]-theophylline nicotinate, 7-[3-(Methyl-2-hydroxyethylamino)-2-hydroxypropyl)]theophylline nicotinate, Angioamin, Angiomin, Complamex, Complamin, Complamin retard, Contamex, Landrina 300, NSC 113217, SK 331A, Sadamine, Stenalgil, Teonicol, Vedrin, Xanthinol niacinate, Xanthinol nicotinate, Xantinol nicotinate, Xavin |
| InChI Formula | InChI=1S/C13H21N5O4.C6H5NO2/c1-15(4-5-19)6-9(20)7-18-8-14-11-10(18)12(21)17(3)13(22)16(11)2;8-6(9)5-2-1-3-7-4-5/h8-9,19-20H,4-7H2,1-3H3;1-4H,(H,8,9) |
| SMILES | CN(CCO)CC(O)Cn1cnc2N(C)C(=O)N(C)C(=O)c12.OC(=O)c3cccnc3 |
| IUPAC Name | 7-[2-hydroxy-3-[2-hydroxyethyl(methyl)amino]propyl]-1,3-dimethylpurine-2,6-dione;pyridine-3-carboxylic acid |
| Show More |