missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Xamoterol hemifumarate

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
β1-adrenoceptor-selective partial agonist (pA2 values are 7.4 - 7.8 and 5.2 - 6.2 at β1- and β2-adrenoceptors respectively).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 397.43 |
| Purity | >97% |
| Molecular Weight (g/mol) | 794.856 |
| Chemical Name or Material | Xamoterol hemifumarate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction