accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 73210-73-8

OHHNNHONOOOHOHHNNHONOOOHOHOOHO(E)

CAS RN 73210-73-8

IUPAC Name: (2E)-but-2-enedioic acid; bis(N-(2-{[2-hydroxy-3-(4-hydroxyphenoxy)propyl]amino}ethyl)morpholine-4-carboxamide)
Synonyms: fumaric acid; bis(xamoterol)
Molecular Weight (g/mol): 794.86
Molecular Formula: C36H54N6O14
InChi Key: QEDVGROSOZBGOZ-WXXKFALUNA-N
SMILES: OC(=O)\C=C\C(O)=O.OC(CNCCNC(=O)N1CCOCC1)COC1=CC=C(O)C=C1.OC(CNCCNC(=O)N1CCOCC1)COC1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results