Learn More
Vortioxetine.HBr, Lipomed
Product Code. 30460837
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 30460837
Brand: Lipomed LPMVOX1884HB10
Please to purchase this item. Need a web account? Register with us today!
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Vortioxetine.HBr
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Vortioxetine Hydrobromide
CAS 960203-27-4
Analyte or Component Names Vortioxetine Hydrobromide
Shipping Condition Room Temperature
Molecular Formula C18 H22 N2 S . Br H
Synonym Piperazine, 1-[2-[(2,4-dimethylphenyl)thio]phenyl]-, hydrobromide (1:1), 1-[2-[(2,4-Dimethylphenyl)thio]phenyl]piperazine hydrobromide, 1-[2-(2,4-Dimethylphenylsulfanyl)phenyl]piperazine hydrobromide, Lu AA 21004 hydrobromide, Vortioxetine hydrobromide
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Br.Cc1ccc(Sc2ccccc2N3CCNCC3)c(C)c1
IUPAC Name 1-[2-(2,4-dimethylphenyl)sulfanylphenyl]piperazine;hydrobromide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 379.358
Quantity 10 mg
Formula Weight 378.077 g/mol