missing translation for 'onlineSavingsMsg'
Learn More

Witeg™ Volumetric Flask with Spout and ST-PE Stopper

Store your chemicals and reagents in conformity certified volumetric flasks. Witeg™Volumetric Flask with Spout and ST-PE Stopper are highly resistant and are labelled with resistant graduation marks.

Brand:  Witeg™ 3 664 025 A

Additional Details : Weight : 0.08800kg

 View more versions of this product

Product Code. 10016301

  • 25.70 € / Each

Estimated Shipment: 27-06-2023
to see stock.
Add to basket

Description

Description

  • Class A.
  • Conformity certified.
  • DIN 12664 (ISO 1042).
  • Borosilicate 3.3.
  • DIFFICO blue acid and alkali resistant ring marks and inscriptions.
  • Spout
  • 10/19 ST-PE stopper
  • Tolerance: 0.040mL.
Specifications

Specifications

Polyethylene
250 mL
Volumetric
Yes
A
ST 14/23
DIFFICO blue acid and alkali resistant ring marks and inscriptions.
Borosilicate Glass 3.3
0.150 mL
Spout
Stopper
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions