Learn More
VOC-Mix 23 6-60 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30325112
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325112 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30325112 Supplier Dr. Ehrenstorfer Supplier No. DREXA05000023ME
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
CAS 75-27-4, 67-66-3, 124-48-1, 127-18-4, 56-23-5, 75-25-2
Quantity 1 mL
Concentration Tetrachloromethane 6 μg/mL∣Tetrachloroethene 20 μg/mL∣Tribromomethane 49.9 μg/mL∣Chloroform 50 μg/mL∣Dibromochloromethane 50 μg/mL∣Bromodichloromethane 50.1 μg/mL∣Trichloroethene 60 μg/mL
Certifications/Compliance ISO/IEC 17025
Solvent or Matrix Methanol