Learn More
Tocris Bioscience™ Vigabatrin
GREENER_CHOICE
Product Code. 18712484
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18712484
Brand: Tocris Bioscience™ 0808/10
Please to purchase this item. Need a web account? Register with us today!
GABA-T inhibitor
Selective GABA-T (transaminase) inhibitor. Anticonvulsant.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 60643-86-9
Molecular Formula C6H11NO2
Specifications
Quantity 10 mg
Formula Weight 129.16
Purity >98%
Molecular Weight (g/mol) 129.159
Chemical Name or Material Vigabatrin