accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 60643-86-9

NH2HOOCH2

CAS RN 60643-86-9

IUPAC Name: 4-aminohex-5-enoic acid
Synonyms: vigabatrin gamma-vinyl gaba 氨己烯酸 GVG vigabatrine vigabatrina vigabatrin hoechst brand yamanouchi brand of vigabatrin gamma-Vinyl-GABA sabrilex
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: PJDFLNIOAUIZSL-UHFFFAOYNA-N
SMILES: NC(CCC(O)=O)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results