Learn More
1,1'-[4,4'Bis(methoxy)biphenyl-3,3'-diyl]bis(tricyclo[3.3.1.13,7]decane), Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
1,1'-[4,4'Bis(methoxy)biphenyl-3,3'-diyl]bis(tricyclo[3.3.1.13,7]decane)
Specifications
Specifications
| Chemical Name or Material | 1,1'-[4,4'Bis(methoxy)biphenyl-3,3'-diyl]bis(tricyclo[3.3.1.13,7]decane) |
| CAS | 932033-57-3 |
| Analyte or Component Names | 1,1'-[4,4'Bis(methoxy)biphenyl-3,3'-diyl]bis(tricyclo[3.3.1.13,7]decane) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C34 H42 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Adapalene Imp. D (EP), Adapalene USP Related Compound D, Adapalene USP RC D, Adapalene Imp. D (Pharmeuropa), 1,1'-[4,4'Bis(methyloxy)-biphenyl-3,3'-diyl]bis(tricyclo[3.3.1.13,7]decane), 1,1'-(4,4'-Dimethoxy[1,1'-biphenyl]-3,3'-diyl)bis-tricyclo[3.3.1.13,7]decane, 3,3'-Diadamantyl-4,4'-dimethoxy-biphenyl, Adapalene Related Compound D, Adapalene Impurity D |
| InChI Formula | InChI=1S/C34H42O2/c1-35-31-5-3-27(13-29(31)33-15-21-7-22(16-33)9-23(8-21)17-33)28-4-6-32(36-2)30(14-28)34-18-24-10-25(19-34)12-26(11-24)20-34/h3-6,13-14,21-26H,7-12,15-20H2,1-2H3 |
| SMILES | COc1ccc(cc1C23CC4CC(CC(C4)C2)C3)c5ccc(OC)c(c5)C67CC8CC(CC(C8)C6)C7 |
| IUPAC Name | 1-[5-[3-(1-adamantyl)-4-methoxyphenyl]-2-methoxyphenyl]adamantane |
| Show More |