Learn More
IBA Lifesciences Strep-Tactin™ Superflow™ High Capacity Column
Product Code. 12806466 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
5 x 0.2 mL
Unit Size:
1mL
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12806466 1 mL 1mL
12856466 5 x 0.2 mL Pack of 5
12826466 5 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12806466 Supplier IBA Lifesciences Supplier No. 21209001
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Gravity flow column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
  • Strep-Tactin™ Superflow™ high capacity columns contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™ and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
  • In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™ Superflow™ high capacity has a higher density of immobilized Strep-Tactin™, which leads to a 3- to 5-fold higher yield of purified protein.
  • To reduce the waiting period during sample and buffer application, the use of the application aid WET FRED is recommended.
Specifications
Quantity 1 mL
For Research Use Only.