missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VGAT Monoclonal Antibody (K40032_6A4), Invitrogen™
Mouse Monoclonal Antibody
480.00 € - 846.00 €
Specifications
| Antigen | VGAT |
|---|---|
| Clone | K40032_6A4 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30200503
|
Invitrogen™
MA556048 |
200 μL |
846.00 €
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30196947
|
Invitrogen™
MA555668 |
50 μL |
480.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: GDEGAEAPVE
VGAT Monoclonal antibody specifically detects VGAT in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| VGAT | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| GABA and glycine transporter; GVAT; hVIAAT; mVGAT; mVIAAT; R75019; rGVAT; SLC32A1; solute carrier family 32 (GABA vesicular transporter), member 1; solute carrier family 32 member 1; vasicular inhibitory amino acid transporter 10D; vesicular GABA transporter; Vesicular inhibitory amino acid transporter; Vgat; Viaat | |
| Human SLC32A1 peptide (immunogen range is 71-80). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SLC32A1 |
| K40032_6A4 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| O35458, Q9H598 | |
| 140679, 83612 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title