missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VGAT Monoclonal Antibody (K40032_6A4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555668
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: GDEGAEAPVE
The Vesicular GABA Amino Acid Transporter (VGAT) is responsible for transport of the inhibitory neurotransmitter into synaptic vesicles (McIntire et al., 1997). The VGAT protein (also known as the Vesicular Inhibitory Amino Aid Transporter or VIAAT) is expressed in synaptic vesicles of both glycine and GABAergic synapses throughout the CNS (Chaudhry et al., 1998). Expression of the VGAT protein changes during development and also in response to patterns of neuronal activity (De et al., 2005).
Specifications
| VGAT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O35458, Q9H598 | |
| SLC32A1 | |
| Human SLC32A1 peptide (immunogen range is 71-80). | |
| 50 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| K40032_6A4 | |
| Unconjugated | |
| SLC32A1 | |
| GABA and glycine transporter; GVAT; hVIAAT; mVGAT; mVIAAT; R75019; rGVAT; SLC32A1; solute carrier family 32 (GABA vesicular transporter), member 1; solute carrier family 32 member 1; vasicular inhibitory amino acid transporter 10D; vesicular GABA transporter; Vesicular inhibitory amino acid transporter; Vgat; Viaat | |
| Mouse | |
| Protein A | |
| RUO | |
| 140679, 83612 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction