Learn More
Verapamil EP Impurity N (as Hydrochloride), Mikromol
Description
Verapamil EP Impurity N (as Hydrochloride), Mikromol
Specifications
Specifications
| Chemical Name or Material | 5,5'-(Methyl-imino)bis[2-(3,4-dimethoxyphenyl)-2-(1-methylethyl)pentanenitrile] Hydrochloride |
| CAS | 2726492-31-3 |
| Analyte or Component Names | 5,5'-(Methyl-imino)bis[2-(3,4-dimethoxyphenyl)-2-(1-methylethyl)pentanenitrile] Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C33 H47 N3 O4 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Verapamil Hydrochloride Imp. N (EP), Verapamil Imp. N (EP), 5,5'-(Methyl-imino)bis[2-(3,4-dimethoxyphenyl)-2-(1-methylethyl)pentanenitrile] Hydrochloride, Verapamil Hydrochloride Imp. N (EP) as Hydrochloride, Verapamil Hydrochloride Impurity N as Hydrochloride, Verapamil Impurity N as Hydrochloride |
| InChI Formula | InChI=1S/C25H31NO3.C2H2O4/c1-4-7-11-23-24(21-10-8-9-12-22(21)29-23)25(27)19-13-15-20(16-14-19)28-18-17-26(5-2)6-3;3-1(4)2(5)6/h8-10,12-16H,4-7,11,17-18H2,1-3H3;(H,3,4)(H,5,6) |
| Solution Type | Neat |
| SMILES | Cl.COc1ccc(cc1OC)C(CCCN(C)CCCC(C#N)(C(C)C)c2ccc(OC)c(OC)c2)(C#N)C(C)C |
| Show More |