missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VASH1 Monoclonal Antibody (K24034_2G6), Invitrogen™
Mouse Monoclonal Antibody
450.00 € - 676.00 €
Specifications
| Antigen | VASH1 |
|---|---|
| Clone | K24034_2G6 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30202817
|
Invitrogen™
MA556054 |
200 μL |
676.00 €
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30193600
|
Invitrogen™
MA555674 |
50 μL |
450.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: WHVLKKVKLG QSVSHDPHSV EQIEWKHSVL DVERLGRDDF RKELERHARD MRLKIGKGTG PPSPTKDRKK DVSSPQRAQS
VASH1 Monoclonal antibody specifically detects VASH1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| VASH1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AI834978; D930046M13Rik; G630009D10Rik; KIAA1036; RGD1564082; TTCP 1; Tubulin carboxypeptidase 1; Tubulinyl-Tyr carboxypeptidase 1; Tyrosine carboxypeptidase 1; VASH; VASH1; vasohibin 1; vasohibin-1 | |
| Recombinant human VASH1 (immunogen range is 251-330). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| VASH1 |
| K24034_2G6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Q7L8A9, Q8C1W1 | |
| 22846, 238328 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title