missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VASH1 Monoclonal Antibody (K24034_2G6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555674
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: WHVLKKVKLG QSVSHDPHSV EQIEWKHSVL DVERLGRDDF RKELERHARD MRLKIGKGTG PPSPTKDRKK DVSSPQRAQS
VASH1 is a protein that possesses actin binding activity and metallocarboxypeptidase activity. It is involved in several processes including negative regulation of angiogenesis, negative regulation of blood vessel endothelial cell migration, and proteolysis. VASH1 acts upstream of or within processes such as negative regulation of endothelial cell migration, negative regulation of endothelial cell proliferation, and negative regulation of lymphangiogenesis. It is located in the apical part of the cell, endoplasmic reticulum, and extracellular space. VASH1 is implicated in liver cirrhosis and portal hypertension, and is a biomarker of liver cirrhosis. Diseases associated with VASH1 include Portal Hypertension and Myasthenic Syndrome, Congenital, 14.
Specifications
| VASH1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q7L8A9, Q8C1W1 | |
| VASH1 | |
| Recombinant human VASH1 (immunogen range is 251-330). | |
| 50 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K24034_2G6 | |
| Unconjugated | |
| VASH1 | |
| AI834978; D930046M13Rik; G630009D10Rik; KIAA1036; RGD1564082; TTCP 1; Tubulin carboxypeptidase 1; Tubulinyl-Tyr carboxypeptidase 1; Tyrosine carboxypeptidase 1; VASH; VASH1; vasohibin 1; vasohibin-1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 22846, 238328 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction