missing translation for 'onlineSavingsMsg'
Learn More
Learn More
US EPA 6020 Internal Standard Stock Solution: 6Li, Sc, Y, In, Tb, Ho, Bi @ 10 μg/mL in 5% HNO3, VHG™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
US EPA 6020 Internal Standard Stock Solution: 6Li, Sc, Y, In, Tb, Ho, Bi @ 10 μg/mL in 5% HNO3
Specifications
Specifications
| Chemical Name or Material | Bismuth/Holmium/Indium/Lithium 6/Scandium/Terbium/Yttrium |
| CAS | 7440-69-9, 7440-60-0, 7440-74-6, 14258-72-1, 7440-20-2, 7440-27-9 |
| Methods | ICP-MS |
| Analyte or Component Names | Bismuth/Holmium/Indium/Lithium 6/Scandium/Terbium/Yttrium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Bismuth 10 μg/mL/Holmium 10 μg/mL/Indium 10 μg/mL/Lithium 6 10 μg/mL/Scandium 10 μg/mL/Terbium 10 μg/mL/Yttrium 10 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction