missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ulex Europaeus Lectin 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 €
Specifications
| Antigen | Ulex Europaeus Lectin 1 |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:500, Immunohistochemistry-Frozen 1:500 |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Ulex Europaeus Lectin 1 Polyclonal specifically detects Ulex Europaeus Lectin 1 in Plant samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Ulex Europaeus Lectin 1 | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Plant | |
| Gorse or Furze Lectin, UEA-1; H(O) lectin 1; Anti H(O) lectin I | |
| IgG | |
| Unpurified | |
| The antibody may be used for studies of the vascular structures. It reacts specifically with UEA-I bound to human endothelial cells of normal and neoplastic blood and lymphatic vessels. Its uniform reaction with vessels enables differentation between vessels and artificial slits in the tissue. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:500, Immunohistochemistry-Frozen 1:500 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Whole antisera, delipidized with 15mM Sodium Azide | |
| Anti-Ulex Europaeus-I (UEA-I) Lectin is developed in rabbit using purified Ulex Europaeus-I lectin from gorse seeds as the immunogen. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title