missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ulex Europaeus Lectin 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-13922-0.05ml
This item is not returnable.
View return policy
Description
Ulex Europaeus Lectin 1 Polyclonal specifically detects Ulex Europaeus Lectin 1 in Plant samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Ulex Europaeus Lectin 1 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:500, Immunohistochemistry-Frozen 1:500 | |
| Gorse or Furze Lectin, UEA-1; H(O) lectin 1; Anti H(O) lectin I | |
| Anti-Ulex Europaeus-I (UEA-I) Lectin is developed in rabbit using purified Ulex Europaeus-I lectin from gorse seeds as the immunogen. | |
| 0.05 mL | |
| Primary | |
| Plant | |
| Antisera |
| Immunohistochemistry | |
| Unconjugated | |
| Whole antisera, delipidized with 15mM Sodium Azide | |
| Rabbit | |
| Unpurified | |
| RUO | |
| The antibody may be used for studies of the vascular structures. It reacts specifically with UEA-I bound to human endothelial cells of normal and neoplastic blood and lymphatic vessels. Its uniform reaction with vessels enables differentation between vessels and artificial slits in the tissue. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction