Description
The tube guide can be placed at different heights to accommodate different length of tubes.
Specifications
Specifications
| Type | Tube Holder |
| Packaging Quantity | 2 Pack |
| Includes | Tube Holder and Guide |
| For Use With | Frac-900 |