missing translation for 'onlineSavingsMsg'
Learn More

Tromethamine, TRC

Product Code. 17949834
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17949834 100 g 100g
30138089 250 g 250g
17919844 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17949834 Supplier Toronto Research Chemicals Supplier No. TRCT892600100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4 H11 N O3
Molecular Weight (g/mol) 121.14
Synonym Trometamol, aminomethylidynetri(methanol), 1,3-Propanediol, 2-amino-2-(hydroxymethyl)-, 2-Amino-1,3-dihydroxy-2-(hydroxymethyl)propane, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-hydroxymethylpropan-1,3-diol, 2-Amino-2-methylol-1,3-propanediol, Addex-Tham, Aminotri(hydroxymethyl)methane, Aminotrimethylolmethane, Aminotris(hydroxymethyl)methane, Methanamine, 1,1,1-tris(hydroxymethyl)-, NSC 103026, NSC 6365, NSC 65434, Pehanorm, TAM, TAM (buffering agent), THAM, Talatrol, Tri Amino, Tri(hydroxymethyl)aminomethane, Tri(hydroxymethyl)methylamine, Trigmo base, Triladyl, Trimethylolaminomethane, Tris, Tris (buffering agent), Tris Amino, Tris Amino Crystal, Tris base, Tris buffer, Tris(hydroxymethyl)aminomethane, Tris(hydroxymethyl)methanamine, Tris(hydroxymethyl)methylamine, Tris(methylolamino)methane, Tris-steril, Trisamin, Trisamine, Trisaminol, Trizma, Trizma Base, Trometamole, Tromethamine, Tromethane, Tromethanmin, Tutofusin tris, [2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amine
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES NC(CO)(CO)CO

Specifications

Quantity 100 g
InChI Formula InChI=1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2
Formula Weight 121.0739
Percent Purity >95%
Chemical Name or Material Trometamol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.