missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris, 99.8 to 100.1% (Dry Basis), Molecular Biology Grade, Ultrapure
Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
Packaging:
Plastic bottle
Unit Size:
10kg
1kg
500g
5kg
Specifications
Specifications
| Melting Point | 171°C to 172°C |
| Boiling Point | 219°C to 220°C (10 mmHg) |
| Physical Form | Crystalline Powder |
| Quantity | 10 kg |
| Linear Formula | NH2C(CH2OH)3 |
| Packaging | Plastic bottle |
| Beilstein | 741883 |
| Formula Weight | 121.14 |
| Percent Purity | 99.8 to 100.1% |
| Grade | Molecular Biology |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction