Learn More
Tris(2,4-di-tert-butylphenyl)phosphite, Mikromol™
Description
Tris(2,4-di-tert-butylphenyl)phosphite
Specifications
Specifications
| Chemical Name or Material | Tris(2,4-di-tert-butylphenyl)phosphite |
| CAS | 31570-04-4 |
| Analyte or Component Names | Tris(2,4-di-tert-butylphenyl)phosphite |
| Shipping Condition | Room Temperature |
| Molecular Formula | C42 H63 O3 P |
| Certifications/Compliance | ISO 9001 |
| InChI Formula | InChI=1S/C42H63O3P/c1-37(2,3)28-19-22-34(31(25-28)40(10,11)12)43-46(44-35-23-20-29(38(4,5)6)26-32(35)41(13,14)15)45-36-24-21-30(39(7,8)9)27-33(36)42(16,17)18/h19-27H,1-18H3 |
| SMILES | CC(C)(C)c1ccc(OP(Oc2ccc(cc2C(C)(C)C)C(C)(C)C)Oc3ccc(cc3C(C)(C)C)C(C)(C)C)c(c1)C(C)(C)C |
| IUPAC Name | tris(2,4-ditert-butylphenyl) phosphite |
| Molecular Weight (g/mol) | 646.92 |
| Show More |