Learn More
Tris(2,4-di-tert-butylphenyl)phosphite 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30325592
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325592 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30325592 Supplier Dr. Ehrenstorfer Supplier No. DREA17894350AL100
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tris(2,4-di-tert-butylphenyl)phosphite
CAS 31570-04-4
Analyte or Component Names Tris(2,4-di-tert-butylphenyl)phosphite
Shipping Condition Room Temperature
Molecular Formula C42 H63 O3 P
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C42H63O3P/c1-37(2,3)28-19-22-34(31(25-28)40(10,11)12)43-46(44-35-23-20-29(38(4,5)6)26-32(35)41(13,14)15)45-36-24-21-30(39(7,8)9)27-33(36)42(16,17)18/h19-27H,1-18H3
SMILES CC(C)(C)c1ccc(OP(Oc2ccc(cc2C(C)(C)C)C(C)(C)C)Oc3ccc(cc3C(C)(C)C)C(C)(C)C)c(c1)C(C)(C)C
IUPAC Name tris(2,4-ditert-butylphenyl) phosphite
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 646.92
Quantity 1 mL
Formula Weight 646.4515 g/mol