Learn More
Trimetazidine N-Oxide Dihydrochloride, Mikromol™
Product Code. 30050072
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050072 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30050072 Supplier Mikromol Supplier No. MM0539.13
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Trimetazidine N-Oxide Dihydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Trimetazidine N-Oxide Dihydrochloride
CAS 2731228-51-4
Analyte or Component Names Trimetazidine N-Oxide Dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C14 H22 N2 O4 . 2 Cl H
Certifications/Compliance ISO 9001
Synonym Trimetazidine N-Oxide Dihydrochloride
InChI Formula InChI=1S/C14H22N2O4.2ClH/c1-18-12-5-4-11(13(19-2)14(12)20-3)10-16(17)8-6-15-7-9-16;;/h4-5,15H,6-10H2,1-3H3;2*1H
SMILES Cl.Cl.COc1ccc(C[N+]2([O-])CCNCC2)c(OC)c1OC
IUPAC Name 1-oxido-1-[(2,3,4-trimethoxyphenyl)methyl]piperazin-1-ium;dihydrochloride
Molecular Weight (g/mol) 355.26
Quantity 100 mg
Formula Weight 354.1113 g/mol