Learn More
Trimeprazine Sulphone N-Oxide Hydrochloride (Oxomemazine N-Oxide Hydrochloride), Mikromol™
Description
Trimeprazine Sulphone N-Oxide Hydrochloride (Oxomemazine N-Oxide Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | Trimeprazine Sulphone N-Oxide Hydrochloride |
| CAS | 2731709-59-2 |
| Analyte or Component Names | Trimeprazine Sulphone N-Oxide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H22 N2 O3 S . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Trimeprazine Sulphone N-Oxide Hydrochloride, Oxomemazine N-Oxide Hydrochloride |
| InChI Formula | InChI=1S/C18H22N2O3S.ClH/c1-14(13-20(2,3)21)12-19-15-8-4-6-10-17(15)24(22,23)18-11-7-5-9-16(18)19;/h4-11,14H,12-13H2,1-3H3;1H |
| SMILES | Cl.CC(CN1c2ccccc2S(=O)(=O)c3ccccc13)C[N+](C)(C)[O-] |
| IUPAC Name | 3-(5,5-dioxophenothiazin-10-yl)-N,N,2-trimethylpropan-1-amine oxide;hydrochloride |
| Show More |