Learn More
Trimeprazine Sulfoxide N-Oxide (Trimeprazine N,S-Dioxide), Mikromol™
Description
Trimeprazine Sulfoxide N-Oxide (Trimeprazine N,S-Dioxide)
Specifications
Specifications
| Chemical Name or Material | Trimeprazine Sulfoxide N-Oxide |
| CAS | 2731414-77-8 |
| Analyte or Component Names | Trimeprazine Sulfoxide N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H22 N2 O2 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-(Methylazomethyl)-N-isopropylbenzamide, Trimeprazine Sulfoxide N-Oxide |
| InChI Formula | InChI=1S/C19H21ClN2O2.2ClH/c20-17-8-6-16(7-9-17)19(15-4-2-1-3-5-15)22-12-10-21(11-13-22)14-18(23)24;;/h1-9,19H,10-14H2,(H,23,24);2*1H |
| SMILES | CC(CN1c2ccccc2S(=O)c3ccccc13)C[N+](C)(C)[O-] |
| IUPAC Name | N,N,2-trimethyl-3-(5-oxophenothiazin-10-yl)propan-1-amine oxide |
| Show More |