Learn More
Trimeprazine N-Oxide Hydrochloride, Mikromol™
Description
Trimeprazine N-Oxide Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Trimeprazine N-Oxide Hydrochloride |
| CAS | 2725579-25-7 |
| Analyte or Component Names | Trimeprazine N-Oxide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H22 N2 O S . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | cis-Paroxetine Hydrochloride, N,N,beta-Trimethyl-10H-phenothiazine-10-propanamine N-Oxide Hydrochloride, Trimeprazine N-Oxide Hydrochloride |
| InChI Formula | InChI=1S/C16H16O2/c1-12(18-2)14-9-6-10-15(11-14)16(17)13-7-4-3-5-8-13/h3-12H,1-2H8 |
| SMILES | Cl.CC(CN1c2ccccc2Sc3ccccc13)C[N+](C)(C)[O-] |
| IUPAC Name | N,N,2-trimethyl-3-phenothiazin-10-ylpropan-1-amine oxide;hydrochloride |
| Show More |