Learn More
Trimebutine N-Oxide Hydrochloride, Mikromol™
Description
Trimebutine N-Oxide Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Trimebutine N-Oxide Hydrochloride |
| CAS | 2731998-25-5 |
| Analyte or Component Names | Trimebutine N-Oxide Hydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C22 H29 N O6 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 6beta-Oxycodol Hydrochloride, Trimebutine N-Oxide Hydrochloride |
| InChI Formula | InChI=1S/C25H30INO3.ClH/c1-4-7-11-23-24(19-10-8-9-12-21(19)30-23)25(28)18-13-14-22(20(26)17-18)29-16-15-27(5-2)6-3;/h8-10,12-14,17H,4-7,11,15-16H2,1-3H3;1H |
| SMILES | Cl.CCC(COC(=O)c1cc(OC)c(OC)c(OC)c1)(c2ccccc2)[N+](C)(C)[O-] |
| IUPAC Name | N,N-dimethyl-2-phenyl-1-(3,4,5-trimethoxybenzoyl)oxybutan-2-amine oxide;hydrochloride |
| Show More |