Learn More
Trimebutine N-Oxide Hydrochloride, Mikromol™
Product Code. 17922346
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17922346 100 mg 100mg
17931846 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17922346 Supplier Mikromol Supplier No. MM0417.17
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Trimebutine N-Oxide Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Trimebutine N-Oxide Hydrochloride
CAS 2731998-25-5
Analyte or Component Names Trimebutine N-Oxide Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C22 H29 N O6 . Cl H
Certifications/Compliance ISO 9001
Synonym 6beta-Oxycodol Hydrochloride, Trimebutine N-Oxide Hydrochloride
InChI Formula InChI=1S/C25H30INO3.ClH/c1-4-7-11-23-24(19-10-8-9-12-21(19)30-23)25(28)18-13-14-22(20(26)17-18)29-16-15-27(5-2)6-3;/h8-10,12-14,17H,4-7,11,15-16H2,1-3H3;1H
SMILES Cl.CCC(COC(=O)c1cc(OC)c(OC)c(OC)c1)(c2ccccc2)[N+](C)(C)[O-]
IUPAC Name N,N-dimethyl-2-phenyl-1-(3,4,5-trimethoxybenzoyl)oxybutan-2-amine oxide;hydrochloride
Molecular Weight (g/mol) 439.93
Quantity 100 mg
Formula Weight 439.1762 g/mol