Learn More
Triflusal, Mikromol™
Product Code. 17922716
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17922716 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17922716 Supplier Mikromol Supplier No. MM3414.00
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Triflusal
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Triflusal
CAS 322-79-2
Analyte or Component Names Triflusal
Shipping Condition Ice pack (-20°C)
Molecular Formula C10 H7 F3 O4
Certifications/Compliance ISO/IEC 17025
Synonym Triflusal, 2-(Acetyloxy)-4-(trifluoromethyl)benzoic acid
InChI Formula InChI=1S/C10H7F3O4/c1-5(14)17-8-4-6(10(11,12)13)2-3-7(8)9(15)16/h2-4H,1H3,(H,15,16)
SMILES CC(=O)Oc1cc(ccc1C(=O)O)C(F)(F)F
IUPAC Name 2-acetyloxy-4-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol) 248.16
Quantity 250 mg
Formula Weight 248.0296 g/mol