Learn More
Trifluoperazine Sulfone N1,N4-Dioxide (Trifluoperazine N1,N4,S,S-Tetraoxide), Mikromol™
Description
Trifluoperazine Sulfone N1,N4-Dioxide (Trifluoperazine N1,N4,S,S-Tetraoxide)
Specifications
Specifications
| Chemical Name or Material | Trifluoperazine Sulfone N1,N4-Dioxide |
| CAS | 2724726-60-5 |
| Analyte or Component Names | Trifluoperazine Sulfone N1,N4-Dioxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H24 F3 N3 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Trifluoperazine Sulfone N1,N4-Dioxide, Trifluoperazine N1,N4,S,S-Tetraoxide |
| InChI Formula | InChI=1S/C21H24F3N3O4S/c1-26(28)11-13-27(29,14-12-26)10-4-9-25-17-5-2-3-6-19(17)32(30,31)20-8-7-16(15-18(20)25)21(22,23)24/h2-3,5-8,15H,4,9-14H2,1H3 |
| SMILES | C[N+]1([O-])CC[N+]([O-])(CCCN2c3ccccc3S(=O)(=O)c4ccc(cc24)C(F)(F)F)CC1 |
| IUPAC Name | 10-[3-(4-methyl-1,4-dioxidopiperazine-1,4-diium-1-yl)propyl]-2-(trifluoromethyl)phenothiazine 5,5-dioxide |
| Show More |