Learn More
Trifluoperazine N4-Oxide Dihydrochloride, Mikromol
Product Code. 30459217
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459217 4 x 25 mg 100mg
30050473 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30459217 Supplier Mikromol Supplier No. MM0441.0800254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Trifluoperazine N4-Oxide Dihydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Trifluoperazine N4-Oxide Dihydrochloride
CAS 2724726-63-8
Analyte or Component Names Trifluoperazine N4-Oxide Dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C21 H24 F3 N3 O S . 2 Cl H
Certifications/Compliance ISO 17025
Synonym Trifluoperazine N4-Oxide Dihydrochloride
InChI Formula InChI=1S/C21H24F3N3OS.2ClH/c1-27(28)13-11-25(12-14-27)9-4-10-26-17-5-2-3-6-19(17)29-20-8-7-16(15-18(20)26)21(22,23)24;;/h2-3,5-8,15H,4,9-14H2,1H3;2*1H
Solution Type Neat
SMILES Cl.Cl.C[N+]1([O-])CCN(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1
IUPAC Name 10-[3-(4-methyl-4-oxidopiperazin-4-ium-1-yl)propyl]-2-(trifluoromethyl)phenothiazine;dihydrochloride
Molecular Weight (g/mol) 496.42
Quantity 4 x 25 mg
Formula Weight 495.1126 g/mol