Learn More
Triethylene Glycol Flufenamate, Mikromol™
Description
Triethylene Glycol Flufenamate
Specifications
Specifications
| Chemical Name or Material | Triethylene Glycol Flufenamate |
| CAS | 30544-48-0 |
| Analyte or Component Names | Triethylene Glycol Flufenamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H22 F3 N O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Triethylene Glycol Flufenamate |
| InChI Formula | InChI=1S/C20H22F3NO5/c21-20(22,23)15-4-3-5-16(14-15)24-18-7-2-1-6-17(18)19(26)29-13-12-28-11-10-27-9-8-25/h1-7,14,24-25H,8-13H2 |
| SMILES | OCCOCCOCCOC(=O)c1ccccc1Nc2cccc(c2)C(F)(F)F |
| IUPAC Name | 2-[2-(2-hydroxyethoxy)ethoxy]ethyl 2-[3-(trifluoromethyl)anilino]benzoate |
| Show More |